missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENN/MADD domain containing 4A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DENND4A.
Brand: Abnova H00010260-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MIEDKGPRVADYFVVAGLTDVSKPLEEEIHFNDACHKVAKPKEPITDVSVIIKSLGEEVPQDYICIDVTPTGLSADLNNGSLVGPQIYLCYRRGRDKPPLTSpecifications
DENN/MADD domain containing 4A | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant DENND4A. | |
DENND4A | |
FLJ33949/IRLB/KIAA0476/MYCPBP | |
Mouse | |
50 μL | |
Yes | |
10260 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_005848 | |
DENND4A | |
DENND4A (NP_005839, 1 a.a. ∼ 101 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |