missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
DP1 Monoclonal Antibody (1DP06 (TFD10)), Invitrogen™
Beskrivning
MA5-11268 targets DP-1 Transcription Factor in WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11268 immunogen is recombinant DP-1 protein.
DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
Specifikationer
Specifikationer
| Antigen | DP1 |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Monoclonal |
| Klona | 1DP06 (TFD10) |
| Koncentration | 0.2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 |
| Gen | TFDP1 |
| Genaccessionsnr. | Q08639, Q14186 |
| Gene Alias | AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?