missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SH2D1B (Tyr127) Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA517932
Additional Details : Weight : 0.01000kg
Description
Phospho-SH2D1B (Tyr127) Polyclonal antibody specifically detects Phospho-SH2D1B (Tyr127) in Human samples. It is validated for Western Blot.Specifications
Phospho-SH2D1B (Tyr127) | |
Polyclonal | |
Unconjugated | |
Sh2d1b | |
Eat2; EAT-2; Eat2a; EAT-2A; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; RGD1563935; SH2 domain containing 1B; SH2 domain containing 1B1; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B; Sh2d1b; Sh2d1b1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
117157 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O14796 | |
Sh2d1b | |
Synthetic peptide sequence (NSNSDpYVDVLP) corresponding to the internal amino acids of SH2D1B | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |