Learn More
EBP50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1090
Additional Details : Weight : 0.01000kg
Description
PA1-090 detects ezrin-radixin-moesin (ERM) binding phosphoprotein of 50 kDa (EBP50) from mouse tissues, human T84 and Calu3 cells as well as recombinant human protein. PA1-090 has been successfully used in Western blot, Immunohistochemistry, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ∽50 kDa protein representing EBP50 from T84 cell extract. Immunohistochemical staining of EBP50 in mouse airway results in staining of the epithelium. The PA1-090 immunogen is recombinant human His-tagged EBP50 obtained from bacteria. All His-tag specific antibody has been removed. This product was epitope affinity purified using a synthetic peptide to residues C R(286) S A S S D T S E E L N S Q D S P(302) of human EBP50.
EBP50 Polyclonal antibody specifically detects EBP50 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
EBP50 | |
Polyclonal | |
Unconjugated | |
SLC9A3R1 | |
EBP50; EBP-50; ERM-binding phosphoprotein; ezrin-radixin-moesin binding phosphoprotein-50; ezrin-radixin-moesin-binding phosphoprotein 50; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; Na+/H+ exchange regulatory co-factor; Nherf; NHE-RF; Nherf1; NHERF-1; NPHLOP2; Regulatory cofactor of Na(+)/H(+) exchanger; SLC9A3 regulator 1; Slc9a3r1; So; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1; solute carrier family 9 isoform A3 regulatory factor 1; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26941, 9368 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O14745, P70441 | |
SLC9A3R1 | |
Bacterially expressed, His-tagged human EBP50. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |