missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDG6 Monoclonal Antibody (1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112681
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is RH7777 cells transfected with EDG6 proteinlysate.
EDG6 Monoclonal antibody specifically detects EDG6 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
EDG6 | |
Monoclonal | |
Unconjugated | |
S1PR4 | |
Edg6; EDG-6; Endothelial differentiation G-protein coupled receptor 6; endothelial differentiation G-protein-coupled receptor 6; endothelial differentiation, G protein coupled receptor 6; endothelial differentiation, G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 6; LPC1; Lysophospholipid receptor C1; S1P receptor 4; S1P receptor Edg-6; S1P(4) receptor; S1p4; S1PR4; SLP4; sphingosine 1-phosphate receptor 4; Sphingosine 1-phosphate receptor Edg-6; sphingosine-1-phosphate receptor 4 | |
Mouse | |
Protein A/G | |
RUO | |
8698 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
1 | |
PBS with 0.08% sodium azide | |
O95977 | |
S1PR4 | |
Synthetic peptide corresponding to the C-terminal of EDG6. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |