missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545183
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LHFCTQTLDV SSNSRTAEKT DKEEEGKVRG ANRGLCQRRL KSRTNKASRL Sequence homology: Human: 100%; Pig: 85%
Endothelin 3 Polyclonal antibody specifically detects Endothelin 3 in Human samples. It is validated for Western Blot.Specifications
Endothelin 3 | |
Polyclonal | |
Unconjugated | |
EDN3 | |
114CH19; EDN3; endothelin 3; endothelin-3; ET 3; Et3; ET-3; HSCR4; lethal spotting; ls; MGC15067; MGC61498; PPET3; Preproendothelin 3; preproendothelin-3; preproET-3; RGD1564825; RP4-614C15.1; tmgc48; WS4B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1908 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P14138 | |
EDN3 | |
Synthetic peptide directed towards the C-terminal of human EDN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |