Learn More
EEF1B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528474
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (85%), Xenopus laevis (84%), Pig (98%), Rabbit (99%), Chicken (100%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EEF1B2 Polyclonal antibody specifically detects EEF1B2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EEF1B2 | |
Polyclonal | |
Unconjugated | |
EEF1B2 | |
2810017J07Rik; EEF1B; EEF1B1; EEF1B2; EF1B; EF-1-beta; Elongation factor 1-beta; eukaryotic translation elongation factor 1 beta 1; eukaryotic translation elongation factor 1 beta 2; eukaryotic translation elongation factor 1-beta homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1933, 55949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O70251, P24534 | |
EEF1B2 | |
Recombinant fragment corresponding to a region within amino acids 33 and 225 of Human EEF1B2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |