missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2K Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517720
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
EEF2K Polyclonal antibody specifically detects EEF2K in Human, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
EEF2K | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O00418, P70531 | |
EEF2K | |
Synthetic peptide corresponding to residues surrounding the N-terminus of human eEF2k | |
100 μL | |
Primary | |
Human, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
EEF2K | |
C86191; Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase; calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; calmodulin-dependent protein kinase III; EC 2.7.11.20; eEF-2 kinase; Eef2k; eEF-2K; EF2K; elongation factor 2 kinase; elongation factor-2 kinase; euk; eukaroytic elongation factor 2 kinase; eukaryotic elongation factor 2 kinase; eukaryotic elongation factor-2 kinase; HSU93850; kinase eEF2K; SMEF2K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25435, 29904 | |
-20°C | |
Liquid |