missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin B2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519361
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
Ephrin B2 Polyclonal antibody specifically detects Ephrin B2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Ephrin B2 | |
Polyclonal | |
Unconjugated | |
Efnb2 | |
EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13642, 1948, 306636 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P52799, P52800 | |
Efnb2 | |
Synthetic peptide sequence (CPKVDSKTVGQYE) corresponding to the internal amino acids of EFNB2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |