missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFL8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542220
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QAGAWVRAVL PVPPEELQPE QVAELWGRGD RIESLSDQVL LLEERLGACS Sequence homology: Cow: 93%; Dog: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%
EGFL8 Polyclonal antibody specifically detects EGFL8 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
EGFL8 | |
Polyclonal | |
Unconjugated | |
EGFL8 | |
C6orf8; EGF like domain multiple 8; EGFL8; EGF-like domain 8; EGF-like domain-containing protein 8; EGF-like protein 8; EGF-like-domain protein 8; EGF-like-domain, multiple 8; Epidermal growth factor-like protein 8; multiple EGF-like domain protein 8; NG3; NG3 protein; UNQ8752/PRO29920; Vascular endothelial statin-2; VE-statin-2 | |
Rabbit | |
Protein A | |
RUO | |
80864 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99944 | |
EGFL8 | |
Synthetic peptide directed towards the C-terminal of human EGFL8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |