Learn More
EGR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527814
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: EGR2 tansfected 293T cell. Predicted reactivity: Mouse (90%), Rat (90%), Xenopus laevis (95%), Pig (95%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EGR2 Polyclonal antibody specifically detects EGR2 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
EGR2 | |
Polyclonal | |
Unconjugated | |
Egr2 | |
AT591; CMT1D; CMT4E; DKFZp686J1957; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; early growth response 2; early growth response protein 2; Egr2; Egr-2; FLJ1454; FLJ14547; Krox20; Krox-20; Krox-20 homolog; KROX-20, Drosophila, homolog (early growth response-2); NGF1-B; OTTHUMP00000062133; Zfp-25; Zfp-6; Zinc finger protein Krox-20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13654, 1959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P08152, P11161 | |
Egr2 | |
Recombinant protein fragment corresponding to a region within amino acids 260 and 451 of Human EGR2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |