Learn More
EHF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540440
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RAAGTAGQLL YSNLQHLKWN GQCSSDLFQS THNVIVKTEQ TEPSIMNTWK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%
EHF Polyclonal antibody specifically detects EHF in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
EHF | |
Polyclonal | |
Unconjugated | |
EHF | |
9030625L19Rik; AU019492; EHF; Epithelium-specific Ets transcription factor 3; ESE3; ESE-3; ESE3 transcription factor; ESE3B; ESEJ; ETS domain-containing transcription factor; ETS homologous factor; hEHF; Unknown (protein for MGC:133971) | |
Rabbit | |
Protein A | |
RUO | |
26298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZC4 | |
EHF | |
Synthetic peptide directed towards the N-terminal of human EHF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |