Learn More
eIF3g Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541775
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SPEPELLPGA PLPPPKEVIN GNIKTVTEYK IDEDGKKFKI VRTFRIETRK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 79%
eIF3g Polyclonal antibody specifically detects eIF3g in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
eIF3g | |
Polyclonal | |
Unconjugated | |
EIF3G | |
44kDa; D0Jmb4; eiF3 delta; eIF3 p42; eIF3 p42 {ECO:0000255; eIF3 p44; eIF3 p44 {ECO:0000255; eIF-3 RNA-binding subunit; eIF-3 RNA-binding subunit {ECO:0000255; eIF-3-delta; eIF3-delta; eIF-3-delta {ECO:0000255; EIF3G; eIF3g {ECO:0000255; Eif3p42; EIF3-P42; eIF3-p44; EIF3S4; eukaryotic translation initiation factor 3 RNA-binding subunit; eukaryotic translation initiation factor 3 RNA-binding subunit {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 4; eukaryotic translation initiation factor 3 subunit 4 {ECO:0000255; eukaryotic translation initiation factor 3 subunit G; eukaryotic translation initiation factor 3 subunit G {ECO:0000255; eukaryotic translation initiation factor 3, subunit 4 (delta); eukaryotic translation initiation factor 3, subunit 4 (delta, 44 kDa); eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD); eukaryotic translation initiation factor 3 subunit p42; eukaryotic translation initiation factor 3, subunit 4 delta; eukaryotic translation | |
Rabbit | |
Protein A | |
RUO | |
8666 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75821 | |
EIF3G | |
Synthetic peptide directed towards the N-terminal of human EIF3S4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |