missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Endothelin B Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528766
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: A375, Rat2. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Pig (100%), Rabbit (100%), Chicken (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifikationer
| Endothelin B Receptor | |
| Polyclonal | |
| Unconjugated | |
| EDNRB | |
| ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1910, 50672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P21451, P24530 | |
| EDNRB | |
| Synthetic peptide corresponding to a region within amino acids 378 and 442 of Human Endothelin B receptor | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering