Learn More
EphB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528680
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (98%), Rat (99%), Xenopus laevis (88%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EphB3 Polyclonal antibody specifically detects EphB3 in Human samples. It is validated for Western Blot.Specifications
EphB3 | |
Polyclonal | |
Unconjugated | |
EPHB3 | |
AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.88 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P54753 | |
EPHB3 | |
Recombinant fragment corresponding to a region within amino acids 607 and 998 of Human EPH receptor B3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |