missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538532
Additional Details : Weight : 0.00900kg
Description
The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).Specifications
Ephrin A2 | |
Polyclonal | |
Unconjugated | |
EFNA2 | |
EPLG6, LERK6, Ephrin A2, EphrinA2, EFNA2, Ephrin-A2, EPH-related receptor tyrosine kinase ligand 6, LERK-6, HEK7 ligand, HEK7-L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13637, 1943 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43921, P52801 | |
EFNA2 | |
A synthetic peptide derived from the internal region of human EFNA2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |