missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 348500
Additional Details : Weight : 0.50000kg
Description
EPHA3 (EPA receptor A3), also known as ETK, HEK, ETK1, HEK4, TYRO4, is belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system (reviewed in 1,2). Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.Specifications
EphA3 | |
Polyclonal | |
Unconjugated | |
P29320 | |
EPHA3 | |
Synthetic peptide derived from the C-terminal region of the human EphA3 receptor protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.25 mg/mL | |
EPHA3 | |
EK4, ETK, ETK1, HEK, HEK4, TYRO4, EPH-like kinase 4, TYRO4 protein tyrosine kinase, eph-like tyrosine kinase 1, ephrin type-A receptor 3, human embryo kinase 1, tyrosine-protein kinase receptor ETK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2042 | |
-20°C | |
Liquid |