missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518586
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
EphB2 Polyclonal antibody specifically detects EphB2 in Human samples. It is validated for Western Blot.Specifications
EphB2 | |
Polyclonal | |
Unconjugated | |
EPHB2 | |
CAPB; cEK5; chicken embryo kinase 5; chicken embryo kinase 5 protein; developmentally-regulated Eph-related tyrosine kinase; DRT; EFL6; EFNB3; EK5; ELK; ELK-related tyrosine kinase; embryo kinase 5 protein CEK5; EPH B2; EPH receptor B2; eph tyrosine kinase 3; EPHB2; EphB2/CTF1; EphB2/CTF2; EPHB3; EPH-like kinase 5; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPHT3; EPLG8; EPTH3; ERK; ETECK; ETK2; HEK2; Hek5; HEK-6; LERK8; MGC87492; NET; Neural kinase; Nuk; Nuk receptor tyrosine kinase; PCBC; Prkm5; protein-tyrosine kinase HEK5; Qek5; Renal carcinoma antigen NY-REN-47; RGD1564232; Sek3; TYRO5; TYRO6; Tyrosine-protein kinase receptor EPH-3; tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P29323 | |
EPHB2 | |
Synthetic peptide sequence (QKDRNHRPKFGQ) corresponding to the internal amino acids of EPHB2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |