Learn More
EphB4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547013
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) EphB6 and rmEphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, rmEphA7, rmEphA8, rmEphB2, and rmEphB3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
EphB4 Polyclonal antibody specifically detects EphB4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Specifications
EphB4 | |
Polyclonal | |
Unconjugated | |
EPHB4 | |
AI042935; Developmental kinase 2; EPH receptor B4; Ephb4; ephrin receptor EphB4; ephrin type-B receptor 4; Hepatoma transmembrane kinase; HTK; Mdk2; mDK-2; MYK1; soluble EPHB4 variant 1; soluble EPHB4 variant 2; soluble EPHB4 variant 3; TYRO11; Tyrosine kinase MYK-1; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase TYRO11 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13846, 2050 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P54760, P54761 | |
EPHB4 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphB4 Leu16-Ala539 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |