Learn More
EGFR Monoclonal Antibody (528), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512872
Additional Details : Weight : 0.01000kg
Description
MA5-12872 targets Epidermal Growth Factor Receptor in FACS and IP applications and shows reactivity with Canine and Human samples. The MA5-12872 immunogen is purified EGFR from A431 cells.
EGFR Monoclonal antibody specifically detects EGFR in Canine, Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
EGFR | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P00533 | |
EGFR | |
Purified EGFR from A431 cells | |
500 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunoprecipitation | |
528 | |
Biotin | |
EGFR | |
2.7.10.1; 9030024J15Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; EC 2.7.10.1; egf receptor; Egfr; EGFR-related peptide; epidermal growth factor receptor; epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); ERBB; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; HER1; kinase EGFR; mENA; NISBD2; Oncogene ERBB; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; Urogastrone; wa2; wa-2; Wa5; waved 2 | |
Mouse | |
Protein A | |
RUO | |
1956, 404306 | |
4° C | |
Liquid |