Learn More
EpCAM Monoclonal Antibody (323/A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512436
3980.50 SEK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA5-12436 targets Epithelial Specific Antigen in FACS, ICC/IF, IHC (P), and RIA applications and shows reactivity with Human and Mouse samples. The MA5-12436 immunogen is mCF-7 human breast cancer cells.
EpCAM Monoclonal antibody specifically detects EpCAM in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA).Specifications
EpCAM | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
P16422, Q99JW5 | |
EPCAM | |
MCF-7 human breast cancer cells | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
323/A3 | |
Unconjugated | |
EPCAM | |
adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
Mouse | |
Protein A/G | |
RUO | |
17075, 4072 | |
4° C | |
Liquid |