missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPRS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542817
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KSEKQNKPQK QNDGQRKDPS KNQGGGLSSS GAGEGQGPKK QTRLGLEAKK Sequence homology: Horse: 79%; Human: 100%; Mouse: 79%; Pig: 83%; Rabbit: 79%; Rat: 79%
EPRS Polyclonal antibody specifically detects EPRS in Human samples. It is validated for Western Blot.Specifications
EPRS | |
Polyclonal | |
Unconjugated | |
EPRS1 | |
2410081F06Rik; 3010002K18Rik; bifunctional aminoacyl-tRNA synthetase; Bifunctional glutamate/proline--tRNA ligase; C79379; Cell proliferation-inducing gene 32 protein; EARS; Eprs; EPRS1; GLNS; GLUPRORS; GluRS; glutamate tRNA ligase; Glutamate--tRNA ligase; Glutamatyl-prolyl-tRNA synthetase; glutamine-proline-tRNA synthetase; glutaminyl-tRNA synthetase; glutamyl-prolyl-tRNA synthetase; glutamyl-prolyl-tRNA synthetase 1; Glutamyl-tRNA synthetase; PARS; PIG32; proliferation-inducing gene 32 protein; proliferation-inducing protein 32; proline tRNA ligase; proline-tRNA ligase; Proline--tRNA ligase; prolyl-tRNA synthetase; ProRS; QARS; QPRS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2058 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P07814 | |
EPRS1 | |
Synthetic peptide directed towards the middle region of human EPRS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |