missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPSTI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543176
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RRLGGSQSET EVRQKQQLQL MQSKYKQKLK REESVRIKKE AEEAELQKMK Sequence homology: Cow: 86%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%
EPSTI1 Polyclonal antibody specifically detects EPSTI1 in Human samples. It is validated for Western Blot.Specifications
EPSTI1 | |
Polyclonal | |
Unconjugated | |
EPSTI1 | |
2310046K10Rik; 5033415K03Rik; BRESI1; epithelial stromal interaction 1; epithelial stromal interaction 1 (breast); epithelial-stromal interaction protein 1; EPSTI1; RGD1563207; Unknown (protein for IMAGE:7987136) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
94240 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96J88 | |
EPSTI1 | |
Synthetic peptide directed towards the N-terminal of human EPSTI1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |