missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO40 Monoclonal Antibody (2E5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA521977
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: QWKKKRYSHG GTSWRVHREI WQFSSLFSKI KSWEFNEVTS MSEHLKSCPF NIVEHKTDPI LLTSMCQPRE QARESLVSTF RIRPRGRYVS
FBXO40 Monoclonal antibody specifically detects FBXO40 in Human samples. It is validated for ELISA.Specifications
FBXO40 | |
Monoclonal | |
0.33 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9UH90 | |
FBXO40 | |
FBXO40 (NP_057382, 620 a.a. ∽ 709 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
2E5 | |
Unconjugated | |
FBXO40 | |
burs-beta; F-box only protein 40; F-box protein 40; FBX40; FBXO40; KIAA1195; Muscle disease-related protein | |
Mouse | |
Affinity chromatography | |
RUO | |
51725 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |