missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Factor IX Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522304
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Predicted reactivity: Mouse (80%), Rat (82%), Dog (87%), Cat (84%), Pig (87%), Rabbit (82%), Rhesus Monkey (96%), Chimpanzee (99%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifikationer
| Factor IX | |
| Polyclonal | |
| Unconjugated | |
| F9 | |
| AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14071, 2158, 24946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P00740, P16294, P16296 | |
| F9 | |
| Recombinant protein encompassing a sequence within the center region of human Factor IX. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering