missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VII Monoclonal Antibody (9031), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA143038
Additional Details : Weight : 0.01000kg
Description
MA1-43038 detects Factor VII in mouse samples. MA1-43038 has successfully been used in Western blot and ELISA applications. MA1-43038 was derived from mouse factor VII.
Factor VII Monoclonal antibody specifically detects Factor VII in Mouse samples. It is validated for ELISA, Western Blot.Specifications
Factor VII | |
Monoclonal | |
Unconjugated | |
F7 | |
AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
Rat | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
14068 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
9031 | |
50% water with 50% glycerol and no preservative | |
P70375 | |
F7 | |
Mouse Factor VII. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |