missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522988
Additional Details : Weight : 0.01000kg
Description
FANCC Polyclonal antibody specifically detects FANCC in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
FANCC | |
Polyclonal | |
Unconjugated | |
FANCC | |
FA3; Fac; Facc; FANCC; Fanconi anemia complementation group C; Fanconi anemia group C protein; Fanconi anemia group C protein homolog; Fanconi anemia, complementation group C; FLJ14675; mir-3074-1; Protein FACC; RP11-80I15.2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Q00597 | |
FANCC | |
N-terminal human FANCC-GST fusion protein. | |
RUO | |
2176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |