Learn More
FANCD2 Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522705
Additional Details : Weight : 0.01000kg
Description
In Western blot, this antibody should recognize a band at ∽166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights. In immunofluorescence, this has been tested in human MMC and IR-treated MEF cells.
FANCD2 Polyclonal antibody specifically detects FANCD2 in Avian, Canine, Human, Marsupial, Mouse, Non-human Primate, Rat, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FANCD2 | |
Polyclonal | |
DyLight 650 | |
FANCD2 | |
2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
211651, 2177, 312641, 394241, 484659 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
50mM sodium borate with no preservative | |
Q6IV68, Q80V62, Q9BXW9 | |
FANCD2 | |
Human FANCD2 fusion protein (N-terminal fragment). | |
50 μL | |
Primary | |
Avian, Canine, Human, Marsupial, Mouse, Non-human Primate, Rat, Zebrafish | |
Antibody | |
IgG |