missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA123348
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa nuclear cell lysate.
FANCC Polyclonal antibody specifically detects FANCC in Human samples. It is validated for Western Blot.Specifications
FANCC | |
Polyclonal | |
Unconjugated | |
FANCC | |
FA3; Fac; Facc; FANCC; Fanconi anemia complementation group C; Fanconi anemia group C protein; Fanconi anemia group C protein homolog; Fanconi anemia, complementation group C; FLJ14675; mir-3074-1; Protein FACC; RP11-80I15.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Q00597 | |
FANCC | |
A synthetic peptide corresponding to the C-terminal 15 amino acids, conjugated to a carrier protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |