missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAS Monoclonal Antibody (H.831.6), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514882
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
FAS Monoclonal antibody specifically detects FAS in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FAS | |
Monoclonal | |
206 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P25445, Q63199 | |
FAS | |
Synthetic peptide corresponding to residues surrounding Lys259 of human Fas, within the intracellular region | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
H.831.6 | |
Unconjugated | |
FAS | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
246097, 355 | |
-20°C | |
Liquid |