Läs mer
F-box protein 11, Mouse, Clone: 4C12, Abnova™
Beskrivning
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | F-box protein 11 |
| Användningsområden | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Klassificering | Monoclonal |
| Klona | 4C12 |
| Konjugera | Unconjugated |
| Beskrivning | Mouse monoclonal antibody raised against a partial recombinant FBXO11. |
| Formulering | PBS with no preservative; pH 7.4 |
| Gen | FBXO11 |
| Genaccessionsnr. | NM_025133 |
| Gene Alias | FBX11/FLJ12673/MGC44383/PRMT9/UBR6/UG063H01/VIT1 |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.