missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528530
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji, Human ESC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the F-box protein family, such as FBXO15, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.Specifications
FBXO15 | |
Polyclonal | |
Unconjugated | |
FBXO15 | |
AU019763; ecat3; F-box only protein 15; F-box protein 15; FBX15; FBXO15; MGC39671 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201456 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8NCQ5 | |
FBXO15 | |
Recombinant fragment corresponding to a region within amino acids 1 and 246 of Human FBXO15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |