Learn More
FBXO8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530512
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji, JC. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (92%), Xenopus laevis (91%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FBXO8 Polyclonal antibody specifically detects FBXO8 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FBXO8 | |
Polyclonal | |
Unconjugated | |
Fbxo8 | |
DC10; F-box only protein 8; F-box protein 8; F-box protein Fbx8; F-box/SEC7 protein FBS; FBS; Fbx8; FBXO8; UNQ1877/PRO4320 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26269, 50753 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NRD0, Q9QZN3 | |
Fbxo8 | |
Recombinant fragment corresponding to a region within amino acids 114 and 319 of Human FBXO8 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |