missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514651
Additional Details : Weight : 0.01000kg
Description
FGFR2 Polyclonal antibody specifically detects FGFR2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
FGFR2 | |
Polyclonal | |
Unconjugated | |
FGFR2 | |
AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor 2; FGF-10; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25 | |
Rabbit | |
Protein A | |
RUO | |
14183, 2263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P21802, P21803 | |
FGFR2 | |
KLH conjugated synthetic peptide between 22-51 amino acids from the N-terminal region of human FGFR2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |