missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL3 Monoclonal Antibody (2C10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518893
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: ESFDCAKCNE SLYGRKYIQT DSGPYCVPCY DNTFANTCAE CQQLIGHDSR ELFYEDRHFH EGCFRCCRCQ RSLADEPFTC QDSELLCNDC YCSAFSSQCS
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.Specifications
FHL3 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q13643 | |
FHL3 | |
FHL3 (NP_004459.2, 3 a.a. ∽ 102 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2C10 | |
Unconjugated | |
FHL3 | |
SLIM2, FHL-3, LIM-only protein FHL3, SLIM-2, four and a half LIM domains protein 3, skeletal muscle LIM-protein 2 | |
Mouse | |
Affinity chromatography | |
RUO | |
2275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |