Learn More
Fibrinogen gamma Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529734
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, human plasma, HepG2 sup., Mouse plasma. Predicted reactivity: Mouse (92%), Rat (91%), Pig (88%), Chicken (81%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fibrinogen gamma Polyclonal antibody specifically detects Fibrinogen gamma in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Fibrinogen gamma | |
Polyclonal | |
Unconjugated | |
Fgg | |
3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2266, 99571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.93 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P02679, Q8VCM7 | |
Fgg | |
Recombinant protein fragment corresponding to a region within amino acids 194 and 368 of Human Fibrinogen gamma | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |