Läs mer
Invitrogen™ Fibroblasts Monoclonal Antibody (ER-TR7)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA140076
Beskrivning
MA1-40076 detects Fibroblasts from mouse samples. MA1-40076 is cross reactive with human tonsil fibroblasts. It does not cross react with purified laminin, collagen types I-V, fibronectin, heparin sulfate proteoglycan,entactin or nidogen. MA1-40076 has been successfully used in immunohistochemistry (frozen and parrafin), flow cytometry, and immunofluorescence applications. Suggested positive control is spleen The MA1-40076 immunogen is mouse thymic stromal cells. Suggested positive control: Spleen; Suggested negative control: Lymphoid cells.
Fibroblasts are the least specialized cells in the connective-tissue family. Thay are dispersed in connective tissue throughout the body, where they secrete a nonrigid extracellular matrix that is rich in type I and/or type III collagen. The activation of fibroblasts by inflammatory stimuli results in their migration, proliferation and deposition of extracellular matrix components, important features involved in both wound healing and fibrosis.
Specifikationer
| Fibroblasts | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Rat | |
| Protein G | |
| RUO | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| ER-TR7 | |
| Unconjugated | |
| fibro-blasts | |
| Mouse thymic stromal cells. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.