missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537163
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 77 kDa. Purity is >95% by SDS-PAGE.
Fibulin 1 Polyclonal antibody specifically detects Fibulin 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Fibulin 1 | |
Polyclonal | |
Unconjugated | |
Fbln1 | |
Basement-membrane protein 90; BM-90; BM-90/fibulin extracellular matrix glyroprotein; extracellular matrix; extracellular matrix and blood protein; FBLN; Fbln1; fbln1c; fbln1d; FIBL1; FIBL-1; fibulin 1; fibulin 1C; fibulin 1D; Fibulin1; fibulin-1; fibulin-1 C varible region; fibulin-1 D; LOW QUALITY PROTEIN: fibulin-1; PP213; wu:fc52c06 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14114, 2192, 315191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P23142, Q08879 | |
Fbln1 | |
A synthetic peptide corresponding to residues in human Fibulin-1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |