Learn More
FOXN4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539174
Additional Details : Weight : 0.00900kg
Description
Members of the winged-helix/forkhead family of transcription factors, such as FOXN4, are characterized by a 110-amino acid DNA-binding domain that can fold into a variant of the helix-turn-helix motif consisting of 3 alpha helices flanked by 2 large loops or wings. These transcription factors are involved in a variety of biologic processes as key regulators in development and metabolism (Li et al., 2004 [PubMed 15363391]).[supplied by OMIM, Mar 2008].Specifications
FOXN4 | |
Polyclonal | |
Unconjugated | |
FOXN4 | |
sli, slipjig | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121643 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96NZ1 | |
FOXN4 | |
A synthetic peptide derived from the internal region of human FOXN4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |