missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR109B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA120430
Additional Details : Weight : 0.01000kg
Description
PA1-20430 detects G Protein-Coupled Receptor HM74 in human samples.
GPR109B Polyclonal antibody specifically detects GPR109B in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GPR109B | |
Polyclonal | |
Unconjugated | |
HCAR3 | |
G protein-coupled receptor 109B; GPR109B; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74B; GTP-binding protein; HCA3; HCAR3; HM74; HM74B; hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; Niacin receptor 2; NIACR2; Nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P49019 | |
HCAR3 | |
Synthetic peptide corresponding to the 3rd cytoplasmic loop of human G Protein-Coupled Receptor HM74. | |
25 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |