missing translation for 'onlineSavingsMsg'
Learn More

G3BP Polyclonal Antibody, Invitrogen™

G3BP Antibody, PA1-27843, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Brand:  Invitrogen PA127843

Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg

Product Code. 11531223

  • 3925.00 SEK / 50µg


Estimated Shipment date
from Supplier 08-04-2019
Add to basket

Description

Description

May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3'UTR. ATP- and magnesium-dependen
Host Species: Chicken
Cross Reactivity: Human, Mouse, Rat
Isotype: IgY
Immunogen: Synthetic peptide corresponding to human Ras-GTPase-activating protein.
Formulation: 50μg purified antibody in PBS and no preservative
Synonyms: G3BP, HDH-VIII, MGC111040
Uniprot Accession: P97855, Q13283
Entrez GeneID: 10146, 171092, 27041
Gene Symbol: G3BP1
PA1-27843 detects G3BP from human, mouse, and rat samples. PA1-27843 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein. The immunogen is a synthetic peptide corresponding to human Ras-GTPase-activating protein.
Specifications

Specifications

G3BP
Polyclonal
50μg purified antibody in PBS and no preservative
Purified
G3BP1
Synthetic peptide corresponding to human Ras-GTPase-activating protein.
50μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
10146, 171092, 27041
Western Blot
Unconjugated
P97855, Q13283
G3BP, HDH-VIII, MGC111040
Chicken
IgY
RUO
Primary
Polyclonal
Human, Murine, Rat
Videos