Läs mer
Invitrogen™ GBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544204
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: HTKGIWMWCV PHPKKPEHTL VLLDTEGLGD IEKGDNENDS WIFALAILLS Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
Specifikationer
| GBP2 | |
| Polyclonal | |
| Unconjugated | |
| GBP2 | |
| GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2634 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P32456 | |
| GBP2 | |
| Synthetic peptide directed towards the N-terminal of human GBP2 (aa 74-123). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |