missing translation for 'onlineSavingsMsg'
Läs mer

anti-GFP Tag FITC, Polyclonal, Invitrogen™

GFP Tag Polyclonal antibody PA1-46326 from Thermo Scientific™ specifically detects GFP Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence and Western Blot.

Brand:  Invitrogen PA1-46326

Produktkod. 11553411

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

GFP Tag Polyclonal antibody PA1-46326 from Thermo Scientific™ specifically detects GFP Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence and Western Blot.

Green fluorescence protein (GFP) is a 27 kDa protein derived from the jellyfish Aequorea victoria, which emits green light (emission peak at a wavelenth of 509 nm) when excited by blue light (excitation peak at a wavelenth of 395 nm). Green Fluorescent Protein (GFP) has become an invaluable tool in cell biology research, since its intrinsic fluorescence can be visualized in living cells. GFP fluorescence is stable under fixation conditions and suitable for a variety of applications. GFP has been widely used as a reporter for gene expression, enabling researchers to visualize and localize GFP-tagged proteins within living cells without the need for chemical staining. Other applications of GFP include assessment of protein protein interactions through the yeast two hybrid system and measurement of distance between proteins through fluorescence energy transfer (FRET) protocols. GFP technnology has considerably contributed to a greater understanding of cellular physiology.
Host Species: Rabbit
Cross Reactivity: Tag
Isotype: IgG
Immunogen: Green fluorescent protein (GFP) fusion protein corresponding to the full length sequence (246 aa) from the jellyfish Aequorea victoria.
Formulation: 50μg Purified Antibody (1mg/ml) in 0.02M potassium phosphate, with 0.15M NaCl, 10mg/ml BSA and 0.01% sodium azide; pH7.2
Synonyms: Green Fluorescence Protein
This antibody shows minimal cross reactivity to human, mouse, and rat serum proteins.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

GFP Tag
Polyclonal
Green Fluorescence Protein
Rabbit
50 μg
Primary
Purified
Tag
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
FITC
gfp
Green fluorescent protein (GFP) fusion protein corresponding to the full length sequence (246 aa) from the jellyfish Aequorea victoria.
RUO
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.