missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525015
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
GLD2 Polyclonal antibody specifically detects GLD2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GLD2 | |
Polyclonal | |
Unconjugated | |
Papd4 | |
8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100715, 167153 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q6PIY7, Q91YI6 | |
Papd4 | |
KLH conjugated synthetic peptide between 58-87 amino acids from the N-terminal region of human GLD2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |