Learn More
GLUT1 Mouse anti-Human, Mouse, Rat, Clone: SPM498, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA137783
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is esophageal or breast carcinoma.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein(Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon,l ung, stomach, esophagus, and breast.Specifications
GLUT1 | |
Monoclonal | |
Unconjugated | |
P11166, P17809, P11167 | |
SLC2A1 | |
Synthetic peptide derived from C-terminus of human GLUT-1 protein | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
SPM498 | |
SLC2A1 | |
Glucose transporter type 1, erythrocyte/brain, HepG2 glucose transporter | |
Mouse | |
Purified | |
RUO | |
6513, 24778, 20525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |