missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA123519
Additional Details : Weight : 0.01000kg
Description
PA1-23519 detects GLUT10 in human samples.
GLUT10 Polyclonal antibody specifically detects GLUT10 in Human samples. It is validated for ELISA, Western Blot.Specifications
GLUT10 | |
Polyclonal | |
Unconjugated | |
SLC2A10 | |
AA450473; ATS; glucose transporter type 10; Glut10; GLUT-10; GLUT11; MGC118830; MGC118833; Slc2a10; SLC2A11-a; SLC2A11-c; solute carrier family 2 (facilitated glucose transporter), member 10; solute carrier family 2 member 10; solute carrier family 2, facilitated glucose transporter member 10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
81031 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O95528 | |
SLC2A10 | |
Human GLUT10 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |