missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Rabbit anti-Bovine, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA116976
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: mouse kidney lysate or protein.
GPI-anchored HDL-binding protein 1 (Gpihbp1) is a novel protein that may be involved in the regulation of the delivery of fats to cells for energy and storage. Digested fats travel to the small intestine, where they are packaged into chylomicrons (particles filled with triglycerides). Chylomicrons then travel through the bloodstream and deliver triglycerides to tissues that are hungry for fuel or to adipose tissue for energy storage. Triglycerides are broken down or hydrolyzed by the enzyme lipoprotein lipase (LpL). The triglyceride breakdown products are then taken up and used by cells. Gpihbp1 is the molecule in capillaries that facilitates the capture of chylomicrons and facilitates the interaction with LpL. It has been shown that fats in the bloodstream are not readily metabolized in the absence of GPIHBP1.Specifications
Gpihbp1 | |
Polyclonal | |
Unconjugated | |
Gpihbp1 | |
GPI-anchored HDL-binding protein 1 antibody, GPI-anchored High-Density Lipid-binding protein 1 antibody, GlycosylPhosphatidylInositol-anchored high-de | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
300027, 512826, 68453 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
tris glycine with 0.15M NaCl and 0.05% sodium azide | |
Q9D1N2 | |
GPIHBP1 | |
Synthetic peptide corresponding to residues 100-170 of mouse GPI-anchored HDL-binding protein 1. | |
100 μL | |
Primary | |
Bovine, Mouse, Rat | |
Antibody | |
IgG |