missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR108 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533602
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Monkey, Rat, Hamster, Dog, Horse (94%) Mouse, Panda, Bovine, Pig (89%).
GPR108 Polyclonal antibody specifically detects GPR108 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR108 | |
Polyclonal | |
Unconjugated | |
GPR108 | |
1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56927 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NPR9 | |
GPR108 | |
Synthetic 18 amino acid peptide from 1st cytoplasmic domain of human GPR108. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |