missing translation for 'onlineSavingsMsg'
Learn More

GPR19 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA533674

Additional Details : Weight : 0.01000kg

Product Code. 15302441

  • 4220.00 SEK / 50µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Elephant, Panda, Bat (100%) Mouse, Bovine, Horse, Rabbit, Stickleback (94%).

GPR19 Polyclonal antibody specifically detects GPR19 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

GPR19
Polyclonal
Unconjugated
GPR19
G protein-coupled receptor 19; GPR19; GPR-NGA; Probable G-protein coupled receptor 19
Rabbit
Antigen affinity chromatography
RUO
2842
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q15760
GPR19
Synthetic 18 amino acid peptide from 2nd cytoplasmic domain of human GPR19.
50 μg
Primary
Human, Non-human Primate
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers