missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533696
Additional Details : Weight : 0.01000kg
Description
GPR32 Polyclonal antibody specifically detects GPR32 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR32 | |
Polyclonal | |
Unconjugated | |
GPR32 | |
G protein-coupled receptor 32; GPCRW; GPR32; Probable G-protein coupled receptor 32; resolvin D1 receptor; RVDR; RVDR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2854 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O75388 | |
GPR32 | |
Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR32. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |